missing translation for 'onlineSavingsMsg'
Learn More

(3-Methylphenyl)(2,4,6-trimethylphenyl)iodonium Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. M29085G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29085G 5 g
M29081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29085G Supplier TCI America Supplier No. M29085G
Only null left

Chemical Identifiers

CAS 197245-87-7
Molecular Formula C17H18F3IO3S
Molecular Weight (g/mol) 486.29
MDL Number MFCD20264883
InChI Key JFTSJYFKINVLMA-UHFFFAOYSA-M
Synonym Mesityl(m-tolyl)iodonium Triflate
PubChem CID 10719708
IUPAC Name (3-methylphenyl)(2,4,6-trimethylphenyl)iodanium trifluoromethanesulfonate
SMILES [O-]S(=O)(=O)C(F)(F)F.CC1=CC([I+]C2=C(C)C=C(C)C=C2C)=CC=C1

Specifications

Melting Point 168°C
Quantity 5 g
Formula Weight 486.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (3-Methylphenyl)(2,4,6-trimethylphenyl)iodonium Trifluoromethanesulfonate