missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylpentane, ≥99.5% (GC), MilliporeSigma™ Supelco™
Supplier: Merck Emd Millipore 6832050ML
Description
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal CareSpecifications
| 96-14-0 | |
| 135 mmHg (17°C) | |
| 0.664 g/mL (at 25°C) | |
| Neat | |
| MFCD00009342 | |
| Analytical Standard | |
| 86.18 | |
| Analytical Standard |
| 532°F | |
| 2.97 (vs. air) | |
| -7°C - closed cup | |
| (CH3 CH2)2 CHCH3 | |
| 64°C | |
| 50 mL | |
| ≥99.5% (GC) |