missing translation for 'onlineSavingsMsg'
Learn More

3-Methylpentane, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187377
Change view
Click to view available options
Quantity:
5 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019815 5 mL
111018172 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019815 Supplier Merck Emd Millipore Supplier No. 683205ML
Only null left
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 96-14-0
Ignition Point 532°F
Vapor Pressure 135 mmHg (17°C)
Vapor Density 2.97 (vs. air)
Density 0.664 g/mL (at 25°C)
Flash Point -7°C - closed cup
Physical Form Neat
Linear Formula (CH3 CH2)2 CHCH3
MDL Number MFCD00009342
Boiling Point 64°C
Type Analytical Standard
Quantity 5 mL
Formula Weight 86.18
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less