accessibility menu, dialog, popup

UserName

CAS RN 96-14-0

IUPAC Name: 3-methylpentane
Synonyms: 3-methylpentane
Molecular Weight (g/mol): 86.18
Molecular Formula: C6H14
InChi Key: PFEOZHBOMNWTJB-UHFFFAOYSA-N
SMILES: CCC(C)CC
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (3)
Grade 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (1)
  • (5)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)