missing translation for 'onlineSavingsMsg'
Learn More

3-Methylcinnamic acid, 98%, Thermo Scientific™

Catalog No. AC296940100
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC296940100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC296940100 Supplier Thermo Scientific Chemicals Supplier No. 296940100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3029-79-6
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00016844
InChI Key JZINNAKNHHQBOS-AATRIKPKSA-N
Synonym 3-methylcinnamic acid, m-methylcinnamic acid, e-3-m-tolyl acrylic acid, 3-m-tolyl acrylic acid, 2e-3-3-methylphenyl acrylic acid, 2e-3-3-methylphenyl prop-2-enoic acid, cinnamic acid, m-methyl, 3-methylcinnamic acid, predominantly trans, 2e-3-3-methylphenyl-2-propenoic acid, 2-propenoic acid, 3-3-methylphenyl-, 2e
PubChem CID 2063448
ChEBI CHEBI:61118
IUPAC Name (E)-3-(3-methylphenyl)prop-2-enoic acid
SMILES CC1=CC=CC(=C1)C=CC(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 116°C to 119°C
Color White
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3C6H4CH=CHCO2H
Quantity 10 g
Formula Weight 162.19
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3-Methylcinnamic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 221-199-2

RUO – Research Use Only