missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbiphenyl 95.0+%, TCI America™

Catalog No. M02965G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M02965G 5 g
M02961G 1 g
M029625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M02965G Supplier TCI America Supplier No. M02965G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 643-93-6
Molecular Formula C13H12
Molecular Weight (g/mol) 168.239
MDL Number MFCD00008533
InChI Key NPDIDUXTRAITDE-UHFFFAOYSA-N
Synonym 3-methylbiphenyl, 3-phenyltoluene, 3-methyl-1,1'biphenyl, 1,1'-biphenyl, 3-methyl, 3-methyl-1,1'-biphenyl, biphenyl, 3-methyl, unii-kfp6eo4j6p, kfp6eo4j6p, 1-methyl-3-phenyl-benzene, 3-methyl-biphenyl
PubChem CID 12564
IUPAC Name 1-methyl-3-phenylbenzene
SMILES CC1=CC=CC(=C1)C2=CC=CC=C2

Specifications

Boiling Point 272°C
Quantity 5 g
Formula Weight 168.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methylbiphenyl
EINECSNumber (4)-0015
TSCA Yes