missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzyl bromide, 97%

Catalog No. AAB2473114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2473114 25 g
AAB2473106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2473114 Supplier Thermo Scientific Chemicals Supplier No. B2473114
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 620-13-3 | C8H9Br | 185.064 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 620-13-3
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000177
InChI Key FWLWTILKTABGKQ-UHFFFAOYSA-N
Synonym 3-methylbenzyl bromide, 1-bromomethyl-3-methylbenzene, alpha-bromo-m-xylene, m-methylbenzyl bromide, m-xylyl bromide, benzene, 1-bromomethyl-3-methyl, 3-bromomethyl toluene, m-xylene, .alpha.-bromo, 3-methylbenzylbromide, m-xylene, alpha-bromo
PubChem CID 12099
IUPAC Name 1-(bromomethyl)-3-methylbenzene
SMILES CC1=CC(=CC=C1)CBr

Specifications

Density 1.37
Boiling Point 184°C to 185°C (340 mmHg)
Flash Point 82°C (179°F)
Refractive Index 1.569
Quantity 25 g
UN Number UN1701
Beilstein 2076425
Merck Index 14,10090
Formula Weight 185.07
Percent Purity 97%
Chemical Name or Material 3-Methylbenzyl bromide
Show More Show Less
Hazard Category H227-H302+H312+H332-H314
Hazard Statement GHS H Statement
H330-H314-H318-H227
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: XYLYL BROMIDE, LIQUID
EINECSNumber 210-625-2
RTECSNumber CY9039200
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only