missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzofuran-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. M13921G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13921G 1 g
M13925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13921G Supplier TCI America Supplier No. M13921G

Chemical Identifiers

CAS 24673-56-1
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.171
MDL Number MFCD00060511
InChI Key YMZTUCZCQMQFMK-UHFFFAOYSA-N
Synonym 3-methylbenzofuran-2-carboxylic acid, 2-benzofurancarboxylic acid, 3-methyl, 3-methyl-2-benzofurancarboxylic acid, 3-methyl-benzofuran-2-carboxylic acid, 3-methylbenzo b furan-2-carboxylic acid, cambridge id 5100668, acmc-1cp97, 3-methylbenzofuran-2-carboxylicacid, 3-methylbenzofuran 2 carboxylic acid, 2-benzofurancarboxylicacid, 3-methyl
PubChem CID 600591
IUPAC Name 3-methyl-1-benzofuran-2-carboxylic acid
SMILES CC1=C(OC2=CC=CC=C12)C(=O)O

Specifications

Melting Point 193°C
Quantity 1 g
Formula Weight 176.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methylbenzofuran-2-carboxylic Acid