missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-phenyl-4-isoxazolecarboxylic acid, 97%, Thermo Scientific™

Catalog No. p-4510227
Change view
Click to view available options
Quantity:
1g
10g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC20301DA 1g Amber Glass Bottle
CC20301EA 10g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC20301DA Supplier Thermo Scientific Chemicals Supplier No. CC20301DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17153-21-8
Molecular Formula C11H9NO3
Molecular Weight (g/mol) 203.197
InChI Key GLNQCTGGLIXRRJ-UHFFFAOYSA-N
Synonym 3-methyl-5-phenylisoxazole-4-carboxylic acid, 3-methyl-5-phenyl-4-isoxazolecarboxylic acid, 4-carboxy-3-methylisoxazol-5-yl benzene, 3-methyl-5-phenyl-4-isoxazole carboxylic acid, 3-methyl-5-phenyl-isoxazole-4-carboxylic acid, 4-isoxazolecarboxylicacid, 3-methyl-5-phenyl, 4-isoxazolecarboxylic acid, 3-methyl-5-phenyl
PubChem CID 2776514
IUPAC Name 3-methyl-5-phenyl-1,2-oxazole-4-carboxylic acid
SMILES CC1=NOC(=C1C(=O)O)C2=CC=CC=C2

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 203.2
Percent Purity 97%
Chemical Name or Material 3-Methyl-5-phenyl-4-isoxazolecarboxylic acid