missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-4-nitropyrazole 98.0+%, TCI America™

Catalog No. M24695G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M24695G Supplier TCI America Supplier No. M24695G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 5334-39-4
Melting Point 135°C
Color White-Yellow
Boiling Point 325°C
Molecular Formula C4H5N3O2
Quantity 5 g
MDL Number MFCD00037864
Synonym 3-methyl-4-nitro-1h-pyrazole, 3-methyl-4-nitropyrazole, 1h-pyrazole, 3-methyl-4-nitro, 3-methyl-4-nitro pyrazole, 3-me-4-no2 pyrazole, 5-methyl-4-nitropyrazole, acmc-209l6a, ksc497o4j, 4-nitro-5-methyl-1h-pyrazole, 3-methyl-4-nitro-1h-pyrazole #
InChI Key WTZYTQJELOHMMJ-UHFFFAOYSA-N
SMILES CC1=C(C=NN1)[N+]([O-])=O
IUPAC Name 5-methyl-4-nitro-1H-pyrazole
Molecular Weight (g/mol) 127.10
PubChem CID 79255
Formula Weight 127.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-4-nitropyrazole
Show More Show Less
RTECSNumber UQ7435000
TSCA No