missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-4-nitrophenol, 98%

Catalog No. AAL0316322
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0316322 100 g
AAL0316314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0316322 Supplier Thermo Scientific Chemicals Supplier No. L0316322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2581-34-2 | C7H7NO3 | 153.14 g/mol

3-Methyl-4-nitrophenol is a biochemical used in the preparation of phosphorothioamidate analogues as antimalarial agents. 3-methyl-4-nitrophenol has anti-androgenic activity. PNMC significantly inhibits basal and hCG-stimulated testosterone production in rats.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-4-nitrophenol is a biochemical used in the preparation of phosphorothioamidate analogues as antimalarial agents. 3-methyl-4-nitrophenol has anti-androgenic activity. PNMC significantly inhibits basal and hCG-stimulated testosterone production in rats.

Solubility
Soluble in water(1.34g/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 2581-34-2
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007333
InChI Key PIIZYNQECPTVEO-UHFFFAOYSA-N
Synonym 4-nitro-m-cresol, 4-nitro-3-cresol, 4-nitro-3-methylphenol, 2-nitro-5-hydroxytoluene, 5-hydroxy-2-nitrotoluene, phenol, 3-methyl-4-nitro, 4-nitro-5-methylphenol, m-cresol, 4-nitro, p-nitro-m-cresol, 4-nitro-1-hydroxy-3-methylbenzene
PubChem CID 17412
ChEBI CHEBI:38683
IUPAC Name 3-methyl-4-nitrophenol
SMILES CC1=CC(O)=CC=C1[N+]([O-])=O

Specifications

Melting Point 126°C to 130°C
Odor Phenol-like
Quantity 100 g
Beilstein 1868105
Solubility Information Soluble in water(1.34g/L).
Formula Weight 153.14
Percent Purity 98%
Chemical Name or Material 3-Methyl-4-nitrophenol
Hazard Category H302-H312-H315-H319-H332-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 219-952-5
RTECSNumber GP2625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only