missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-buten-1-ol 98.0+%, TCI America™

Catalog No. M072625ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M072625ML 25 mL
M0726100ML 100 mL
M0726500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M072625ML Supplier TCI America Supplier No. M072625ML

Chemical Identifiers

CAS 763-32-6
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00002933
InChI Key CPJRRXSHAYUTGL-UHFFFAOYSA-N
Synonym 3-methyl-3-buten-1-ol, isobutenylcarbinol, 3-buten-1-ol, 3-methyl, isoprenol, methallylcarbinol, 3-isopentenyl alcohol, 2-methyl-1-buten-4-ol, isopropenylethyl alcohol, methallyl carbinol, 3-methyl-3-butenol
PubChem CID 12988
ChEBI CHEBI:62898
IUPAC Name 3-methylbut-3-en-1-ol
SMILES CC(=C)CCO

Specifications

Color Yellow
Boiling Point 130°C
Quantity 25 mL
UN Number 1987
Formula Weight 86.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-3-buten-1-ol
EINECSNumber (2)-2393
RTECSNumber EM9473100
TSCA Yes