missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-buten-1-ol, 97%

Catalog No. AC156631000
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156631000 100 mL Glass bottle
AC156635000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC156631000 Supplier Thermo Scientific Chemicals Supplier No. 156631000

CAS: 763-32-6 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 763-32-6
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00002933
InChI Key CPJRRXSHAYUTGL-UHFFFAOYSA-N
Synonym 3-methyl-3-buten-1-ol, isobutenylcarbinol, 3-buten-1-ol, 3-methyl, isoprenol, methallylcarbinol, 3-isopentenyl alcohol, 2-methyl-1-buten-4-ol, isopropenylethyl alcohol, methallyl carbinol, 3-methyl-3-butenol
PubChem CID 12988
ChEBI CHEBI:62898
IUPAC Name 3-methylbut-3-en-1-ol
SMILES CC(=C)CCO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8500g/mL
Boiling Point 132°C
Flash Point 36°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4327 to 1.4347
Linear Formula H2C=C(CH3)CH2CH2OH
Quantity 100 mL
Specific Gravity 0.85
Solubility Information Solubility in water: 170g/L (20 c)
Formula Weight 86.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Methyl-3-buten-1-ol, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-110-8
TSCA TSCA

RUO – Research Use Only