missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-3-buten-1-ol, 97%

Catalog No. AAB23398AE
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB23398AE 100 mL
AAB23398AC 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB23398AE Supplier Thermo Scientific Chemicals Supplier No. B23398AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 763-32-6 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 763-32-6
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00002933
InChI Key CPJRRXSHAYUTGL-UHFFFAOYSA-N
Synonym 3-methyl-3-buten-1-ol, isobutenylcarbinol, 3-buten-1-ol, 3-methyl, isoprenol, methallylcarbinol, 3-isopentenyl alcohol, 2-methyl-1-buten-4-ol, isopropenylethyl alcohol, methallyl carbinol, 3-methyl-3-butenol
PubChem CID 12988
ChEBI CHEBI:62898
IUPAC Name 3-methylbut-3-en-1-ol
SMILES CC(=C)CCO

Specifications

Density 0.85
Boiling Point 130°C to 133°C
Flash Point 36°C (97°F)
Refractive Index 1.433
Quantity 100 mL
UN Number UN1987
Beilstein 1071239
Formula Weight 86.13
Percent Purity 97%
Chemical Name or Material 3-Methyl-3-buten-1-ol
Hazard Category H226-H319
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 212-110-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only