missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-pentanone, 98+%

Catalog No. AAA1999918
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1999918 50 g
AAA1999909 10 g
AAA1999914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1999918 Supplier Thermo Scientific Chemicals Supplier No. A1999918
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 565-61-7 | C6H12O | 100.161 g/mol

3-Methyl-2-pentanone is used as a solvent and as an intermediate for syntheses of pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-2-pentanone is used as a solvent and as an intermediate for syntheses of pharmaceuticals.

Solubility
Soluble in alcohol.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container.

Chemical Identifiers

CAS 565-61-7
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00009336
InChI Key UIHCLUNTQKBZGK-UHFFFAOYSA-N
Synonym 3-methyl-2-pentanone, 2-pentanone, 3-methyl, methyl sec-butyl ketone, sec-butyl methyl ketone, methyl 1-methylpropyl ketone, +/--3-methyl-pentan-2-one, methylethylacetone, sec-c4h9coch3, 3-methyl-pentan-2-one, acmc-1alw4
PubChem CID 11262
IUPAC Name 3-methylpentan-2-one
SMILES CCC(C)C(=O)C

Specifications

Density 0.815
Boiling Point 118°C to 119°C
Flash Point 12°C (53°F)
Refractive Index 1.4 00
Quantity 50 g
UN Number UN1224
Beilstein 635791
Solubility Information Soluble in alcohol.
Formula Weight 100.16
Percent Purity ≥98%
Chemical Name or Material 3-Methyl-2-pentanone
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 209-282-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only