missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. M137025G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M137025G 25 g
M1370250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M137025G Supplier TCI America Supplier No. M137025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5437-38-7
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.15
MDL Number MFCD00007180
InChI Key DGDAVTPQCQXLGU-UHFFFAOYSA-N
Synonym 3-methyl-2-nitrobenzoic acid, 2-nitro-m-toluic acid, 2-nitro-3-methylbenzoic acid, benzoic acid, 3-methyl-2-nitro, unii-61wop984ab, 3-methyl-2-nitro-benzoic acid, 3-methyl-2-nitrobenzoicacid, pubchem8352, acmc-1ashh, dsstox_cid_5640
PubChem CID 21575
IUPAC Name 3-methyl-2-nitrobenzoic acid
SMILES CC1=CC=CC(C(O)=O)=C1[N+]([O-])=O

Specifications

Melting Point 223°C
Color White
Quantity 25 g
Formula Weight 181.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-2-nitrobenzoic Acid