missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-cyclopentenone 98.0+%, TCI America™

Catalog No. M095625ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M095625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M095625ML Supplier TCI America Supplier No. M095625ML

Chemical Identifiers

CAS 2758-18-1
Molecular Formula C6H8O
Molecular Weight (g/mol) 96.129
MDL Number MFCD00001403
InChI Key CHCCBPDEADMNCI-UHFFFAOYSA-N
Synonym 3-methyl-2-cyclopenten-1-one, 3-methylcyclopent-2-enone, 2-cyclopenten-1-one, 3-methyl, 3-methyl-2-cyclopentenone, 1-methyl-1-cyclopenten-3-one, unii-6v7rsw7273, fema no. 3435, 3-methylcyclopentenone, acmc-1crym, 3-methyl-2cyclopenten-1-one
PubChem CID 17691
IUPAC Name 3-methylcyclopent-2-en-1-one
SMILES CC1=CC(=O)CC1

Specifications

Color Yellow
Boiling Point 108°C
Quantity 25 mL
Formula Weight 96.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-2-cyclopentenone
EINECSNumber (3)-4463
TSCA Yes