missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-cyclohexen-1-one 98.0+%, TCI America™

Catalog No. M10435G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10435G 5 g
M104325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M10435G Supplier TCI America Supplier No. M10435G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1193-18-6
Molecular Formula C7H10O
Molecular Weight (g/mol) 110.16
MDL Number MFCD00001581
InChI Key IITQJMYAYSNIMI-UHFFFAOYSA-N
Synonym 3-methyl-2-cyclohexen-1-one, seudenone, 3-methyl-2-cyclohexenone, 2-cyclohexen-1-one, 3-methyl, methylcyclohexenone, caswell no. 561ab, fema no. 3360, 3-methyl-2-cyclohexene-1-one, epa pesticide chemical code 219700, ghl.pd_mitscher_leg0.699
PubChem CID 14511
IUPAC Name 3-methylcyclohex-2-en-1-one
SMILES CC1=CC(=O)CCC1

Specifications

Color Yellow
Boiling Point 197°C
Quantity 5 g
Formula Weight 110.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methyl-2-cyclohexen-1-one
RTECSNumber GW7340000
TSCA Yes