missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-cyclohexen-1-one, 98%

Catalog No. AAA1570422
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1570422 100 g
AAA1570409 10 g
AAA1570414 25 g
AAA1570418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1570422 Supplier Thermo Scientific Chemicals Supplier No. A1570422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1193-18-6 | C7H10O | 110.16 g/mol

3-Methyl-2-cyclohexen-1-one was used in the synthesis of 19-nor-1α, 25-dihydroxyvitamin D(3) derivatives. It was also used in sex pheromone of the Douglas-fir beetle. Used in nut flavor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-2-cyclohexen-1-one was used in the synthesis of 19-nor-1α, 25-dihydroxyvitamin D(3) derivatives. It was also used in sex pheromone of the Douglas-fir beetle. Used in nut flavor.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away form oxidizing agent.

Chemical Identifiers

CAS 1193-18-6
Molecular Formula C7H10O
Molecular Weight (g/mol) 110.16
MDL Number MFCD00001581
InChI Key IITQJMYAYSNIMI-UHFFFAOYSA-N
Synonym 3-methyl-2-cyclohexen-1-one, seudenone, 3-methyl-2-cyclohexenone, 2-cyclohexen-1-one, 3-methyl, methylcyclohexenone, caswell no. 561ab, fema no. 3360, 3-methyl-2-cyclohexene-1-one, epa pesticide chemical code 219700, ghl.pd_mitscher_leg0.699
PubChem CID 14511
IUPAC Name 3-methylcyclohex-2-en-1-one
SMILES CC1=CC(=O)CCC1

Specifications

Density 0.971
Boiling Point 199°C to 200°C
Flash Point 68°C (154°F)
Odor Pleasant
Refractive Index 1.494
Quantity 100 g
Beilstein 1560601
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 110.15
Percent Purity 98%
Chemical Name or Material 3-Methyl-2-cyclohexen-1-one
Show More Show Less
Hazard Category H227-H302
Hazard Statement GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.
Precautionary Statement P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c
EINECSNumber 214-769-7
RTECSNumber GW7340000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only