missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-benzothiazolinonehydrazone Hydrochloride Hydrate 98.0+%, TCI America™

Catalog No. M21555G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M21555G 5 g
M21551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M21555G Supplier TCI America Supplier No. M21555G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 149022-15-1
Molecular Formula C8H10ClN3S
Molecular Weight (g/mol) 215.70
MDL Number MFCD00149370
InChI Key OEZPVSPULCMUQB-UHFFFAOYSA-N
Synonym unii-9u2c4w7epr, mbth hydrochloride, 9u2c4w7epr, 3-methyl-2-benzothiazolone hydrazone hydrochloride, 3-methyl-2-benzothiazolinone hydrazone hydrochloride, z-2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride, mbth hcl, ccris 1543, 2 3h-benzothiazolone, 3-methyl-, hydrazone, hydrochloride 1:1, 2e-2-hydrazinylidene-3-methyl-1,3-benzothiazole hydrochloride
PubChem CID 9575839
IUPAC Name 2-hydrazinylidene-3-methyl-2,3-dihydro-1,3-benzothiazole hydrochloride
SMILES Cl.CN1C(SC2=CC=CC=C12)=NN

Specifications

Quantity 5 g
UN Number 2811
Formula Weight 215.70
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-2-benzothiazolinonehydrazone Hydrochloride Hydrate [for Biochemical Research]
RTECSNumber DL7160200
TSCA No