missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-benzothiazolinone Hydrazone Hydrochloride TS, 0.1% (w/v) in 90% (v/v) Methanol, Ricca Chemical

Catalog No. 483816
Change view
Click to view available options
Package: Natural Poly; Size: 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
483816 Package: Natural Poly; Size: 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 483816 Supplier Ricca Chemical Company Supplier No. 483816

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Natural Poly Bottle
CAS 149022-15-1, 67-56-1, 7732-18-5
Concentration or Composition Notes 0.1% (w/v)
CAS Min % 0.12, 86.0, 12.11
CAS Max % 0.13, 89.0, 12.61
Molecular Formula CH4O
MDL Number MFCD00004595
Synonym unii-9u2c4w7epr, mbth hydrochloride, 9u2c4w7epr, 3-methyl-2-benzothiazolone hydrazone hydrochloride, 3-methyl-2-benzothiazolinone hydrazone hydrochloride, z-2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride, mbth hcl, ccris 1543, 2 3h-benzothiazolone, 3-methyl-, hydrazone, hydrochloride 1:1, 2e-2-hydrazinylidene-3-methyl-1,3-benzothiazole hydrochloride
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
Grade Test Solution
Chemical Name or Material 3-Methyl-2-benzothiazolinone Hydrazone Hydrochloride
Show More Show Less