missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-benzothiazolinone Hydrazone Hydrochloride TS, 0.1% (w/v) in 90% (v/v) Methanol, Ricca Chemical

Catalog No. 48384
Change view
Click to view available options
Quantity:
120 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
48384 120 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48384 Supplier Ricca Chemical Company Supplier No. 48384

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 149022-15-1 , 67-56-1 , 7732-18-5
Molecular Formula C8H10ClN3S
Molecular Weight (g/mol) 215.70
MDL Number MFCD00004595
InChI Key OEZPVSPULCMUQB-UHFFFAOYSA-N
Synonym unii-9u2c4w7epr, mbth hydrochloride, 9u2c4w7epr, 3-methyl-2-benzothiazolone hydrazone hydrochloride, 3-methyl-2-benzothiazolinone hydrazone hydrochloride, z-2-hydrazono-3-methyl-2,3-dihydrobenzo d thiazole hydrochloride, mbth hcl, ccris 1543, 2 3h-benzothiazolone, 3-methyl-, hydrazone, hydrochloride 1:1, 2e-2-hydrazinylidene-3-methyl-1,3-benzothiazole hydrochloride
IUPAC Name 2-hydrazinylidene-3-methyl-2,3-dihydro-1,3-benzothiazole hydrochloride
SMILES Cl.CN1C(SC2=CC=CC=C12)=NN

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 0.1% (w/v)
CAS Min % 0.12, 86.0, 12.11
CAS Max % 0.13, 89.0, 12.61
Physical Form Liquid
Color Colorless
Solvent or Matrix 90% (v/v) Methanol
Quantity 120 mL
Grade Test Solution
Chemical Name or Material 3-Methyl-2-benzothiazolinone Hydrazone Hydrochloride