missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2,4-pentanedione, mixture of tautomers, 95%

Catalog No. AAL0113514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0113514 25 g
AAL0113506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0113514 Supplier Thermo Scientific Chemicals Supplier No. L0113514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 815-57-6 | C6H10O2 | 114.144 g/mol

3-Methyl-2,4-pentanedione involves in asymmetric substitution of 1,3-diphenyl-2-propenyl acetate catalyzed by amphiphilic resin-supported monodentate phosphine ligands can occur.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-2,4-pentanedione involves in asymmetric substitution of 1,3-diphenyl-2-propenyl acetate catalyzed by amphiphilic resin-supported monodentate phosphine ligands can occur.

Solubility
Soluble in water.

Notes
Air & heat sensitive. Do not store together with strong bases and oxidizing agents.

Chemical Identifiers

CAS 815-57-6
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00008762
InChI Key GSOHKPVFCOWKPU-UHFFFAOYSA-N
Synonym 3-methyl-2,4-pentanedione, methylacetylacetone, 1,1-diacetylethane, 2,4-pentanedione, 3-methyl, methyl acetylacetone, 3-acetylbutan-2-one, 3-methyl-acetylacetone, acmc-209plp, 3-methyl-2,4-pentandione, 3-methylpentan-2,4-dione
PubChem CID 69949
IUPAC Name 3-methylpentane-2,4-dione
SMILES CC(C(=O)C)C(=O)C

Specifications

Density 0.98
Boiling Point 170°C to 171°C
Flash Point 56°C (132°F)
Refractive Index 1.442
Quantity 25 g
UN Number UN1224
Beilstein 635909
Solubility Information Soluble in water.
Formula Weight 114.14
Percent Purity 95%
Chemical Name or Material 3-Methyl-2,4-pentanedione, mixture of tautomers
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
EINECSNumber 212-420-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only