missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene 98.0+%, TCI America™

Catalog No. M23915G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23915G 5 g
M23911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23915G Supplier TCI America Supplier No. M23915G

Chemical Identifiers

CAS 885692-91-1
Molecular Formula C11H17BO2S
Molecular Weight (g/mol) 224.125
MDL Number MFCD08063137
InChI Key DGJCULPYCMEHFZ-UHFFFAOYSA-N
Synonym 2-(3-Methyl-2-thienyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-Methyl-2-thiopheneboronic Acid Pinacol Ester
PubChem CID 45588125
IUPAC Name 4,4,5,5-tetramethyl-2-(3-methylthiophen-2-yl)-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C=CS2)C

Specifications

Melting Point 60°C
Color White-Yellow
Quantity 5 g
Formula Weight 224.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene
TSCA No
Recommended Storage Refrigerator