missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-1-phenyl-2-phospholene 1-oxide, 94%

Catalog No. AAA1179222
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1179222 100 g
AAA1179206 5 g
AAA1179214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1179222 Supplier Thermo Scientific Chemicals Supplier No. A1179222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 707-61-9 | C11H13OP | 192.198 g/mol

3-Methyl-1-phenyl-2-phospholene 1-oxide is used as a catalyst for intramolecular aza-wittig cyclization reactions and polymerization reactions. It is a reactant used for the preparation of phospha sugars and their radical bromination derivatives as anticancer agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-1-phenyl-2-phospholene 1-oxide is used as a catalyst for intramolecular aza-wittig cyclization reactions and polymerization reactions. It is a reactant used for the preparation of phospha sugars and their radical bromination derivatives as anticancer agents.

Solubility
Soluble in polar solvents.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 707-61-9
Molecular Formula C11H13OP
Molecular Weight (g/mol) 192.198
MDL Number MFCD00014518
InChI Key YMKWWHFRGALXLE-UHFFFAOYSA-N
Synonym 3-methyl-1-phenyl-2-phospholene 1-oxide, 3-methyl-1-phenyl-2-phospholene-1-oxide, 2,3-dihydro-4-methyl-1-phenyl-1h-phosphole 1-oxide, 1h-phosphole, 2,3-dihydro-4-methyl-1-phenyl-, 1-oxide, 4-methyl-1-phenyl-2,3-dihydro-1h-phosphole 1-oxide, 2-phospholene, 3-methyl-1-phenyl-, 1-oxide, 2,3-dihydro-4-methyl-1-phenyl-1h-phosphole, 1-oxide, 4-methyl-1-phenyl-2,3-dihydro-1$l^ 5-phosphole 1-oxide, 3-methyl-1-phenyl-4,5-dihydro-1??-phosphol-1-one, 2-phospholene, 1-oxide
PubChem CID 69722
IUPAC Name 4-methyl-1-phenyl-2,3-dihydro-1$l^{5}-phosphole 1-oxide
SMILES CC1=CP(=O)(CC1)C2=CC=CC=C2

Specifications

Melting Point 60°C to 65°C
Boiling Point 173°C to 174°C (0.7 mmHg)
Odor Unpleasant
Quantity 100 g
Beilstein 609223
Sensitivity Hygroscopic
Solubility Information Soluble in polar solvents.
Formula Weight 192.19
Percent Purity 94%
Chemical Name or Material 3-Methyl-1-phenyl-2-phospholene 1-oxide
Hazard Category Warning
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 211-901-5
RTECSNumber SZ6105100
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only