missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-1-hexanol 97.0+%, TCI America™
Spécifications
Spécifications
| Color | Colorless |
| Boiling Point | 96°C |
| Quantity | 5 g |
| Formula Weight | 116.20 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-Methyl-1-hexanol |
Sécurité et manipulation
| EINECSNumber | (2)-0217 |
| TSCA | No |