missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Méthyl-1-butanol 99,0+%, TCI America™
Spécifications
Spécifications
| Color | Incolore |
| Boiling Point | 132°C |
| Quantity | 25 mL |
| UN Number | 1105 |
| Formula Weight | 88.15 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | 3-Methyl-1-butanol |
Sécurité et manipulation
| EINECSNumber | (2)-0217 |
| RTECSNumber | EL5425000 |
| TSCA | Yes |