missing translation for 'onlineSavingsMsg'
Learn More

3-Méthoxypyridine 96,0+%, TCI America™

Numéro de catalogue. M23045G
Modifier l'affichage
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
M23045G 5 g
M23041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. M23045G Fournisseur TCI America Code fournisseur M23045G

Identifiants chimiques

CAS 7295-76-3
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00673022
InChI Key UMJSCPRVCHMLSP-UHFFFAOYSA-N
Synonym pyridine, 3-methoxy, beta-methoxypyridine, 3-methyoxypyridine, unii-xv2a2d8595, .beta.-methoxypyridine, 3-methoxy pyridine, 3-methoxypiridine, 3-methoxy-pyridine, pubchem15370, acmc-209oov
PubChem CID 23719
IUPAC Name 3-méthoxypyridine
SMILES COC1=CN=CC=C1

Spécifications

Color Jaune
Boiling Point 179°C
Quantity 5 g
Formula Weight 109.13
Percent Purity ≥96.0% (GC,T)
Physical Form Liquide
Chemical Name or Material 3-Methoxypyridine