missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxypyridine, 97%

Catalog No. AC393080250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC393080250 25 g Glass bottle
AC393080050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC393080250 Supplier Thermo Scientific Chemicals Supplier No. 393080250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7295-76-3 | C6H7NO | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7295-76-3
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00673022
InChI Key UMJSCPRVCHMLSP-UHFFFAOYSA-N
Synonym pyridine, 3-methoxy, beta-methoxypyridine, 3-methyoxypyridine, unii-xv2a2d8595, .beta.-methoxypyridine, 3-methoxy pyridine, 3-methoxypiridine, 3-methoxy-pyridine, pubchem15370, acmc-209oov
PubChem CID 23719
IUPAC Name 3-methoxypyridine
SMILES COC1=CN=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0830g/mL
Boiling Point 65°C (15.0 mmHg)
Flash Point 70°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5190 to 1.5220 (20°C, 589nm)
Quantity 25 g
Specific Gravity 1.083
Formula Weight 109.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Methoxypyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 230-730-7

RUO – Research Use Only