missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxypropylamine, 99+%, Thermo Scientific Chemicals

Catalog No. AC257520050
Change view
Click to view available options
Quantity:
5 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC257520050 5 mL Glass bottle
AC257520010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC257520050 Supplier Thermo Scientific Chemicals Supplier No. 257520050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5332-73-0 | Molecular Formula: C4H11NO | Molecular Weight: 89.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5332-73-0
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.14
MDL Number MFCD00014831
InChI Key FAXDZWQIWUSWJH-UHFFFAOYSA-N
Synonym 3-methoxypropylamine, 1-propanamine, 3-methoxy, 1-amino-3-methoxypropane, 3-aminopropyl methyl ether, 3-methyoxypropylamine, 3-methoxy-n-propylamine, 3-methoxy-1-propanamine, propylamine, 3-methoxy, 3-mpa, propanolamine methyl ether
PubChem CID 1672
IUPAC Name 3-methoxypropan-1-amine
SMILES COCCCN

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -65°C
Color Colorless
Density 0.8700g/mL
Boiling Point 118°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4165 to 1.4185
Linear Formula CH3OCH2CH2CH2NH2
Quantity 5 mL
Specific Gravity 0.87
Solubility Information Solubility in water: miscible. Other solubilities: miscible with ethanol,toluene,acetone,ether,,carbon tetrachloride and hexane
Viscosity 0.91 mPa.s (20°C)
Formula Weight 89.12
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 3-Methoxypropylamine, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Flammable liquid and vapor.
May cause an allergic skin reaction.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 226-241-3
RTECSNumber UI3335000
TSCA TSCA

RUO – Research Use Only