missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methoxypropionitrile 99,0+%, TCI America™
Spécifications
Spécifications
| Melting Point | -63°C |
| Color | Jaune |
| Boiling Point | 166°C |
| Quantity | 25 mL |
| Formula Weight | 85.11 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | 3-Methoxypropionitrile |
Sécurité et manipulation
| EINECSNumber | (2)-1536 |
| RTECSNumber | TZ4870000 |
| TSCA | Yes |