missing translation for 'onlineSavingsMsg'
Learn More

3'-Methoxybiphenyl-4-sulfonyl chloride, 95%, Thermo Scientific™

Catalog No. AC450480050
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450480050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC450480050 Supplier Thermo Scientific Chemicals Supplier No. 450480050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 186550-26-5
Molecular Formula C13H11ClO3S
Molecular Weight (g/mol) 282.75
InChI Key DVQBUJTYQXKLKZ-UHFFFAOYSA-N
Synonym 3'-methoxy-biphenyl-4-sulfonyl chloride, 3'-methoxybiphenyl-4-sulfonyl chloride, 3'-methoxy 1,1'-biphenyl-4-sulfonyl chloride, 3'-methoxy-1,1'-biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 3'-methoxy, 4-3-methoxyphenyl benzenesulfonyl chloride, 3'-methoxy-biphenyl-4-sulfonylchloride, 3/'-methoxybiphenyl-4-sulfonyl chloride
PubChem CID 10826798
IUPAC Name 4-(3-methoxyphenyl)benzenesulfonyl chloride
SMILES COC1=CC=CC(=C1)C2=CC=C(C=C2)S(=O)(=O)Cl

Specifications

Assay Percent Range 94% min. (HPLC)
Quantity 5g
Formula Weight 282.75
Percent Purity 95%
Chemical Name or Material 3'-Methoxybiphenyl-4-sulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

RUO – Research Use Only