missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzylamine, 97%

CAS: 5071-96-5 | C8H11NO | 137.18 g/mol

Supplier:  Thermo Scientific Chemicals L0205414

 View more versions of this product

Catalog No. AAL0205414

Only null left
Explore available promotions

Description

Description

3-Methoxybenzylamine is used in the preparation of 6-substituted purines

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxybenzylamine is used in the preparation of 6-substituted purines

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

3-Methoxybenzylamine
1.066
>110°C (230°F)
1.547
25 g
1447182
3-methoxybenzylamine, m-methoxybenzylamine, 3-methoxyphenyl methanamine, 1-3-methoxyphenyl methanamine, benzenemethanamine, 3-methoxy, 3-methoxyphenyl methylamine, benzylamine, m-methoxy, methoxybenzylamine, 3, benzenemethanamine, 3-methoxy-9ci, sz4
GRRIMVWABNHKBX-UHFFFAOYSA-N
(3-methoxyphenyl)methanamine
21156
≥98%
5071-96-5
230°C to 231°C
C8H11NO
MFCD00008115
UN2735
Air Sensitive
Slightly soluble in water.
COC1=CC=CC(CN)=C1
137.18
137.18

Chemical Identifiers

5071-96-5
137.18
GRRIMVWABNHKBX-UHFFFAOYSA-N
21156
COC1=CC=CC(CN)=C1
C8H11NO
MFCD00008115
3-methoxybenzylamine, m-methoxybenzylamine, 3-methoxyphenyl methanamine, 1-3-methoxyphenyl methanamine, benzenemethanamine, 3-methoxy, 3-methoxyphenyl methylamine, benzylamine, m-methoxy, methoxybenzylamine, 3, benzenemethanamine, 3-methoxy-9ci, sz4
(3-methoxyphenyl)methanamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312+H332-H314

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 225-779-6

RTECSNumber : DP5550000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only