missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzylamine, 97%

Catalog No. p-7043682
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0205406 5 g
AAL0205414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0205406 Supplier Thermo Scientific Chemicals Supplier No. L0205406
Only null left

CAS: 5071-96-5 | C8H11NO | 137.18 g/mol

3-Methoxybenzylamine is used in the preparation of 6-substituted purines

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxybenzylamine is used in the preparation of 6-substituted purines

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 5071-96-5
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00008115
InChI Key GRRIMVWABNHKBX-UHFFFAOYSA-N
Synonym 3-methoxybenzylamine, m-methoxybenzylamine, 3-methoxyphenyl methanamine, 1-3-methoxyphenyl methanamine, benzenemethanamine, 3-methoxy, 3-methoxyphenyl methylamine, benzylamine, m-methoxy, methoxybenzylamine, 3, benzenemethanamine, 3-methoxy-9ci, sz4
PubChem CID 21156
IUPAC Name (3-methoxyphenyl)methanamine
SMILES COC1=CC=CC(CN)=C1

Specifications

Density 1.066
Boiling Point 230°C to 231°C
Flash Point >110°C (230°F)
Refractive Index 1.547
Quantity 5 g
UN Number UN2735
Beilstein 1447182
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 137.18
Percent Purity ≥98%
Chemical Name or Material 3-Methoxybenzylamine
Show More Show Less
Hazard Category H302+H312+H332-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 225-779-6
RTECSNumber DP5550000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only