accessibility menu, dialog, popup

UserName

CAS RN 5071-96-5

IUPAC Name: 1-(3-methoxyphenyl)methanamine
Synonyms: 1-(3-methoxyphenyl)methanamine
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: GRRIMVWABNHKBX-UHFFFAOYSA-N
SMILES: COC1=CC=CC(CN)=C1
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)