missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methoxybenzyl Chloride 96.0+%, TCI America™
Supplier: TCI America M082025G
Specifications
| 3-Methoxybenzyl Chloride | |
| Yellow | |
| C8H9ClO | |
| 25 g | |
| 3-methoxybenzyl chloride, 1-chloromethyl-3-methoxybenzene, m-methoxybenzyl chloride, m-chloromethyl anisole, benzene, 1-chloromethyl-3-methoxy, anisole, m-chloromethyl, 3-methoxybenzylchloride, 3-chloromethyl anisole, 3-methoxy benzyl chloride, benzyl chloride,3-methoxy | |
| COC1=CC=CC(CCl)=C1 | |
| 156.61 | |
| 156.61 | |
| Liquid |
| 824-98-6 | |
| 125°C | |
| MFCD00000907 | |
| 3265 | |
| VGISFWWEOGVMED-UHFFFAOYSA-N | |
| 1-(chloromethyl)-3-methoxybenzene | |
| 69994 | |
| ≥96.0% (GC) |
Chemical Identifiers
| 824-98-6 | |
| 156.61 | |
| VGISFWWEOGVMED-UHFFFAOYSA-N | |
| 69994 | |
| COC1=CC=CC(CCl)=C1 |
| C8H9ClO | |
| MFCD00000907 | |
| 3-methoxybenzyl chloride, 1-chloromethyl-3-methoxybenzene, m-methoxybenzyl chloride, m-chloromethyl anisole, benzene, 1-chloromethyl-3-methoxy, anisole, m-chloromethyl, 3-methoxybenzylchloride, 3-chloromethyl anisole, 3-methoxy benzyl chloride, benzyl chloride,3-methoxy | |
| 1-(chloromethyl)-3-methoxybenzene |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator