missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzyl Chloride 96.0+%, TCI America™

Catalog No. p-7132383
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
M08205G 5 g
M082025G 25 g
2 options

Catalog No. M08205G

Supplier: TCI America M08205G

Only null left

Chemical Identifiers

CAS 824-98-6
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.61
MDL Number MFCD00000907
InChI Key VGISFWWEOGVMED-UHFFFAOYSA-N
Synonym 3-methoxybenzyl chloride, 1-chloromethyl-3-methoxybenzene, m-methoxybenzyl chloride, m-chloromethyl anisole, benzene, 1-chloromethyl-3-methoxy, anisole, m-chloromethyl, 3-methoxybenzylchloride, 3-chloromethyl anisole, 3-methoxy benzyl chloride, benzyl chloride,3-methoxy
PubChem CID 69994
IUPAC Name 1-(chloromethyl)-3-methoxybenzene
SMILES COC1=CC=CC(CCl)=C1

Specifications

Color Yellow
Boiling Point 125°C
Quantity 5 g
UN Number 3265
Formula Weight 156.61
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methoxybenzyl Chloride
TSCA No
Recommended Storage Refrigerator