accessibility menu, dialog, popup

UserName

CAS RN 824-98-6

IUPAC Name: 1-(chloromethyl)-3-methoxybenzene
Synonyms: benzyl chloride,3-methoxy
Molecular Weight (g/mol): 156.61
Molecular Formula: C8H9ClO
InChi Key: VGISFWWEOGVMED-UHFFFAOYSA-N
SMILES: COC1=CC=CC(CCl)=C1
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)