missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxybenzoic acid, 99%

Catalog No. p-7080992
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1076718 50 g
AAA1076730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1076718 Supplier Thermo Scientific Chemicals Supplier No. A1076718

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 586-38-9 | C8H8O3 | 152.149 g/mol

3-Methoxybenzoic acid is used in the synthesis and characterization of 3-methoxybenzoates of europium (III) and gadolinium (III)1. It was used in conversion of aromatic carboxylic acids into methyl esters and reduction to the corresponding primary alcohols using a sodium borohydride-THF-methanol system

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxybenzoic acid is used in the synthesis and characterization of 3-methoxybenzoates of europium (III) and gadolinium (III)1. It was used in conversion of aromatic carboxylic acids into methyl esters and reduction to the corresponding primary alcohols using a sodium borohydride-THF-methanol system

Solubility
Soluble in 95% ethanol (50 mg/ml), water (2 mg/ml at 25°C) and methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.

Chemical Identifiers

CAS 586-38-9
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00002499
InChI Key XHQZJYCNDZAGLW-UHFFFAOYSA-N
Synonym m-anisic acid, m-methoxybenzoic acid, benzoic acid, 3-methoxy, 3-anisic acid, meta-anisic acid, 3-methoxy-benzoic acid, unii-e2i36fh6qz, m-methylsalicylic acid, e2i36fh6qz, chembl22425
PubChem CID 11461
IUPAC Name 3-methoxybenzoic acid
SMILES COC1=CC=CC(=C1)C(=O)O

Specifications

Melting Point 104°C to 108°C
Boiling Point 170°C to 172°C (10 mmHg)
Odor Odorless
Quantity 50 g
Beilstein 508838
Solubility Information Soluble in 95% ethanol (50mg/ml),water (2mg/ml at 25°C) and methanol.
Formula Weight 152.15
Percent Purity 99%
Chemical Name or Material 3-Methoxybenzoic acid
Hazard Category H302-H315-H319
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
EINECSNumber 209-574-9
RTECSNumber BZ4375000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only