missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-5-methylphenol 98.0+%, TCI America™

Catalog No. M08955G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08955G 5 g
M08951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08955G Supplier TCI America Supplier No. M08955G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3209-13-0
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00059261
InChI Key NOTCZLKDULMKBR-UHFFFAOYSA-N
Synonym 3-hydroxy-5-methoxytoluene, phenol, 3-methoxy-5-methyl, 5-methoxy-m-cresol, orcinol monomethyl ether, o-methylorcinol, 3-methoxy-5-methyl-phenol, unii-uya9w3l847, 5-methoxy-3-methylphenol, orcinyl ter, 3-methoxy-5-methylphenol
PubChem CID 76674
IUPAC Name 3-methoxy-5-methylphenol
SMILES CC1=CC(=CC(=C1)OC)O

Specifications

Melting Point 63°C
Color White-Yellow
Quantity 5 g
Formula Weight 138.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methoxy-5-methylphenol