missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-5-methylphenol, 97%

Catalog No. AAH2751603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2751603 1 g
AAH2751606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2751603 Supplier Thermo Scientific Chemicals Supplier No. H2751603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3209-13-0 | C8H10O2 | 138.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3209-13-0
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00059261
InChI Key NOTCZLKDULMKBR-UHFFFAOYSA-N
Synonym 3-hydroxy-5-methoxytoluene, phenol, 3-methoxy-5-methyl, 5-methoxy-m-cresol, orcinol monomethyl ether, o-methylorcinol, 3-methoxy-5-methyl-phenol, unii-uya9w3l847, 5-methoxy-3-methylphenol, orcinyl ter, 3-methoxy-5-methylphenol
PubChem CID 76674
IUPAC Name 3-methoxy-5-methylphenol
SMILES CC1=CC(=CC(=C1)OC)O

Specifications

Melting Point 60°C to 66°C
Quantity 1 g
Formula Weight 138.17
Percent Purity 97%
Chemical Name or Material 3-Methoxy-5-methylphenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 221-716-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only