missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-4-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. M20835G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20835G 5 g
M208325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20835G Supplier TCI America Supplier No. M20835G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5081-36-7
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD00007353
InChI Key PWURRRRGLCVBMX-UHFFFAOYSA-N
Synonym 3-methoxy-4-nitrobenzoic acid, 4-nitro-3-methoxybenzoic acid, 4-nitro-m-anisic acid, benzoic acid, 3-methoxy-4-nitro, 3-methoxy-4-nitrobenzoicacid, pubchem3918, acmc-1bn9s, ksc270m6t, 3-methoxy4-nitro-benzoic acid, 3-methoxy-4-nitro-benzoic acid
PubChem CID 78764
SMILES COC1=C(C=CC(=C1)C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 235°C
Quantity 5 g
Formula Weight 197.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methoxy-4-nitrobenzoic Acid