missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-2-methylbenzoic Acid 98.0+%, TCI America™

Catalog No. M22175G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22175G 5 g
M221725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22175G Supplier TCI America Supplier No. M22175G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 55289-06-0
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD02094039
InChI Key JPCISVSOTKMFPG-UHFFFAOYSA-N
Synonym 3-methoxy-o-toluic acid, 2-methyl-m-anisic acid, 3-methoxy-2-methyl-benzoic acid, 3-methoxy-2-methylbenzoicacid, benzoic acid, 3-methoxy-2-methyl, 2-methyl-3-methoxybenzoic acid, pubchem4976, acmc-209lm1, ksc269c9h, 2-methyl-3-methoxy-benzoic acid
PubChem CID 7021483
IUPAC Name 3-methoxy-2-methylbenzoic acid
SMILES COC1=CC=CC(C(O)=O)=C1C

Specifications

Melting Point 149°C
Color White-Yellow
Quantity 5 g
Formula Weight 166.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methoxy-2-methylbenzoic Acid