missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-2-methylbenzoic acid, 97%

Catalog No. AAL1872918
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1872918 50 g
AAL1872909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1872918 Supplier Thermo Scientific Chemicals Supplier No. L1872918
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 55289-06-0 | C9H10O3 | 166.18 g/mol

3-Methoxy-2-methylbenzoic acid is used as a Corey-Bakshi-Shibata oxazaborolidine catalyst for asymmetric reduction and asymmetric synthesis. It is used in the asymmetric reduction of prochiral ketones. Other applications include the enantioselective synthesis of α-hydroxy acids, α-amino acids, C2 symmetrical ferrocenyl diols and propargyl alcohols. It is also used in desymmetrizing reduction leading to (S)-4-hydroxycyclohexenone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxy-2-methylbenzoic acid is used as a Corey-Bakshi-Shibata oxazaborolidine catalyst for asymmetric reduction and asymmetric synthesis. It is used in the asymmetric reduction of prochiral ketones. Other applications include the enantioselective synthesis of α-hydroxy acids, α-amino acids, C2 symmetrical ferrocenyl diols and propargyl alcohols. It is also used in desymmetrizing reduction leading to (S)-4-hydroxycyclohexenone.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 55289-06-0
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD02094039
InChI Key JPCISVSOTKMFPG-UHFFFAOYSA-N
Synonym 3-methoxy-o-toluic acid, 2-methyl-m-anisic acid, 3-methoxy-2-methyl-benzoic acid, 3-methoxy-2-methylbenzoicacid, benzoic acid, 3-methoxy-2-methyl, 2-methyl-3-methoxybenzoic acid, pubchem4976, acmc-209lm1, ksc269c9h, 2-methyl-3-methoxy-benzoic acid
PubChem CID 7021483
IUPAC Name 3-methoxy-2-methylbenzoic acid
SMILES COC1=CC=CC(C(O)=O)=C1C

Specifications

Quantity 50 g
Beilstein 510395
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 166.18
Percent Purity 97%
Chemical Name or Material 3-Methoxy-2-methylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only