missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-2(1H)-pyridone, 97%

Catalog No. AC163540050
Change view
Click to view available options
Quantité:
1 g
5 g
Emballage:
Bouteille en verre
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité Emballage
AC163540050 5 g Bouteille en verre
AC163540010 1 g Bouteille en verre
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC163540050 Supplier Thermo Scientific Chemicals Supplier No. 163540050
Obtenez jusqu’à 1,800 $ en produits chimiques
View all promotions

missing translation for 'validMainframeMsg'

CAS : 20928-63-6 | C6H7NO2 | 125,13 g/mol

Ce produit de marque Thermo Scientific Chemicals faisait à l’origine partie du portefeuille Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit/article (SKU) original d’Acros Organics n’a pas changé dans le cadre de la transition vers la marque Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20928-63-6
Formule moléculaire C6H7NO2
Poids moléculaire (g/mol) 125.13
Numéro MDL MFCD00006015
Clé InChI LKIMDXQLHFCXQF-UHFFFAOYSA-N
Synonyme 3-methoxypyridin-2-ol, 2-hydroxy-3-methoxypyridine, 3-methoxy-2 1h-pyridone, 2 1h-pyridinone, 3-methoxy, 3-methoxypyridin-2 1h-one, 3-methoxypyridin-2-one, 3-methoxy-2-pyridone, 2-pyridinol, 3-methoxy, 3-methoxy-2-hydroxypyridine, methyloxypyridon
PubChem CID 88731
Nom de l’IUPAC 3-méthoxy-1H-pyridine-2-one
SOURIRES COC1=CC=CNC1=O

Specifications

Nom ou substance chimique 3-Methoxy-2(1H)-pyridone
CAS % minimum 96.0
CAS % maximal 100.0
Point de fusion 113,0 °C à 115,0 °C
Couleur Blanc à brun
Spectre infrarouge Authentique
Fourchette de pourcentages d’essai 96% min. (HPLC)
Emballage Bouteille en verre
Quantité 5 g
Poids de formule 125.13
Pourcentage de pureté 97%
Forme physique Poudre ou poudre cristalline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 244-116-1

RUO – Usage réservé à la recherche