missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-1,2-propanediol 98.0+%, TCI America™

Catalog No. M012525ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M012525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M012525ML Supplier TCI America Supplier No. M012525ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 623-39-2
Color Yellow
Boiling Point 220°C
Molecular Formula C4H10O3
Quantity 25 mL
MDL Number MFCD00004719
Synonym 3-methoxy-1,2-propanediol, 1,2-propanediol, 3-methoxy, 1-o-methylglycerol, glycerol 1-monomethyl ether, glycerin-alpha-monomethyl ether, glycerin-.alpha.-monomethyl ether, glycerin methyl ether, 3-methoxypropan-1,2-diol, 1-o-methyl-rac-glycerol, acmc-1bhm7
InChI Key PSJBSUHYCGQTHZ-UHFFFAOYNA-N
SMILES COCC(O)CO
IUPAC Name 3-methoxypropane-1,2-diol
Molecular Weight (g/mol) 106.12
PubChem CID 92822
Formula Weight 106.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Methoxy-1,2-propanediol
Show More Show Less
RTECSNumber TY8170000
TSCA Yes