missing translation for 'onlineSavingsMsg'
Learn More

3-Mercapto-2-pentanone 97.0+%, TCI America™

Catalog No. p-7132976
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M20265G 5 g
M202625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M20265G Supplier TCI America Supplier No. M20265G

Chemical Identifiers

CAS 67633-97-0
Molecular Formula C5H10OS
Molecular Weight (g/mol) 118.194
MDL Number MFCD02093290
InChI Key SZECUQRKLXRGSJ-UHFFFAOYSA-N
Synonym 3-mercapto-2-pentanone, 2-pentanone, 3-mercapto, 3-mercaptopentan-2-one, fema no. 3300, 3-mercapto-2-penanone, 2-pentanone,3-mercapto, acmc-1b6ys, 2-pentanone, 3-mercapto-7ci,9ci
PubChem CID 62237
IUPAC Name 3-sulfanylpentan-2-one
SMILES CCC(C(=O)C)S

Specifications

Color Yellow
Boiling Point 52°C
Quantity 5 g
UN Number 3336
Formula Weight 118.19
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Mercapto-2-pentanone