missing translation for 'onlineSavingsMsg'
Learn More

3-Mercapto-1-hexanol, 96%

Catalog No. AC347560250
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC347560250 25 mL Glass bottle
AC347560050 5 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC347560250 Supplier Thermo Scientific Chemicals Supplier No. 347560250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 51755-83-0 | C6H14OS | 134.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 51755-83-0
Molecular Formula C6H14OS
Molecular Weight (g/mol) 134.23
MDL Number MFCD00792515
InChI Key TYZFMFVWHZKYSE-UHFFFAOYSA-N
Synonym 3-mercapto-1-hexanol, 3-mercaptohexanol, 3-mercaptohexan-1-ol, 1-hexanol, 3-mercapto, 3-thiohexan-1-ol, 3-sulphanylhexan-1-ol, 3-sulfanyl-1-hexanol, 3-thiohexanol, acmc-209kvm, 3-mercapto-hexan-1-ol
PubChem CID 521348
ChEBI CHEBI:77690
IUPAC Name 3-sulfanylhexan-1-ol
SMILES CCCC(CCO)S

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.9700g/mL
Boiling Point 250°C
Flash Point >100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H3C(CH2)2CH(SH)(CH2)2OH
Refractive Index 1.4790 to 1.481
Quantity 25 mL
Specific Gravity 0.97
Solubility Information Solubility in water: insoluble in water.,soluble in organic solvents (most)
Formula Weight 134.23
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3-Mercapto-1-hexanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only