missing translation for 'onlineSavingsMsg'
Learn More

3-Maleimidopropionic Acid 97.0+%, TCI America™

Catalog No. M19625G
Change view
Click to view available options
Quantity:
200 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19625G 5 g
M1962200MG 200 mg
M19621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M19625G Supplier TCI America Supplier No. M19625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7423-55-4
Molecular Formula C7H7NO4
Molecular Weight (g/mol) 169.14
MDL Number MFCD00043030
InChI Key IUTPJBLLJJNPAJ-UHFFFAOYSA-N
Synonym 3-maleimidopropionic acid, 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl propanoic acid, n-maleoyl-beta-alanine, 3-2,5-dioxopyrrol-1-yl propanoic acid, 1h-pyrrole-1-propanoic acid, 2,5-dihydro-2,5-dioxo, maleimide-ch2 2-cooh, n-carboxyethylmaleimide, 3-maleimdepropionicacid, pubchem11845, acmc-209otw
PubChem CID 573621
IUPAC Name 3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanoic acid
SMILES OC(=O)CCN1C(=O)C=CC1=O

Specifications

Melting Point 110°C
Color White
Quantity 5 g
Formula Weight 169.14
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Maleimidopropionic Acid