missing translation for 'onlineSavingsMsg'
Learn More

3-Isopropoxy-4-(tri-n-butylstannyl)-3-cyclobutene-1,2-dione, 97%

Catalog No. AAH51488MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH51488MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAH51488MD Supplier Thermo Scientific Chemicals Supplier No. H51488MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 129034-70-4 | C19H34O3Sn | 429.188 g/mol

3-Isopropoxy-4-(tri-n-butylstannyl)-3-cyclobutene-1,2-dione is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Isopropoxy-4-(tri-n-butylstannyl)-3-cyclobutene-1,2-dione is used as pharmaceutical intermediate.

Solubility
Sparingly soluble in water.

Notes
Stable under recommended storage conditions. Store aw

Chemical Identifiers

CAS 129034-70-4
Molecular Formula C19H34O3Sn
Molecular Weight (g/mol) 429.188
MDL Number MFCD01319011
InChI Key AMVLEBHYELFWJT-UHFFFAOYSA-N
Synonym 3-isopropoxy-4-tributylstannyl-1,2-cyclobutenedione, 3-isopropoxy-4-tributylstannyl cyclobut-3-ene-1,2-dione, 3-cyclobutene-1,2-dione, 3-1-methylethoxy-4-tributylstannyl, acmc-20dn4e, 3-isopropoxy-4-tributylstannyl-cyclobut-3-ene-1,2-dione, 3-isopropoxy-4-tributylstannylcyclobut-3-ene-1,2-dione, 3,4-dioxo-2-isopropoxycyclobut-1-en-1-yl tributylstannane, 3-isopropoxy-4-tri-n-butyl-stannyl-3-cyclobutene-1,2-dione, 3-isopropoxy-4-tri-n-butylstannyl-3-cyclobutene-1,2-dione
PubChem CID 2736683
IUPAC Name 3-propan-2-yloxy-4-tributylstannylcyclobut-3-ene-1,2-dione
SMILES CCCC[Sn](CCCC)(CCCC)C1=C(C(=O)C1=O)OC(C)C

Specifications

Quantity 250 mg
UN Number UN2788
Solubility Information Sparingly soluble in water.
Formula Weight 429.17
Percent Purity 97%
Chemical Name or Material 3-Isopropoxy-4-(tri-n-butylstannyl)-3-cyclobutene-1,2-dione
Hazard Category H301-H312-H315-H319-H372
Hazard Statement GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only